missing translation for 'onlineSavingsMsg'
Learn More

Bisoprolol Hemifumarate 98.0+%, TCI America™

Catalog No. B3994200MG
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B3994200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B3994200MG Supplier TCI America Supplier No. B3994200MG
Only null left

Chemical Identifiers

CAS 104344-23-2
Molecular Formula C40H66N2O12
Molecular Weight (g/mol) 766.97
MDL Number MFCD00792743,MFCD00792743,MFCD00792743,MFCD00865795
InChI Key VMDFASMUILANOL-WXXKFALUNA-N
Synonym bisoprolol fumarate, 1-4-2-isopropoxyethoxy methyl phenoxy-3-isopropylamino propan-2-ol fumarate, but-2-enedioic acid; 1-propan-2-ylamino-3-4-2-propan-2-yloxyethoxymethyl phenoxy-2-propanol, but-2-enedioic acid; 1-propan-2-ylamino-3-4-2-propan-2-yloxyethoxymethyl phenoxy propan-2-ol
PubChem CID 53394791
IUPAC Name (2E)-but-2-enedioic acid; bis(1-[(propan-2-yl)amino]-3-(4-{[2-(propan-2-yloxy)ethoxy]methyl}phenoxy)propan-2-ol)
SMILES OC(=O)\C=C\C(O)=O.CC(C)NCC(O)COC1=CC=C(COCCOC(C)C)C=C1.CC(C)NCC(O)COC1=CC=C(COCCOC(C)C)C=C1

Specifications

Melting Point 102°C
Color White
Quantity 200 mg
Formula Weight 383.49
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Bisoprolol Hemifumarate
RTECSNumber UB8390000
TSCA No