missing translation for 'onlineSavingsMsg'
Learn More

Bisoprolol, ≥98% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111015118
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015118 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015118 Supplier Merck Emd Millipore Supplier No. 5078725MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 66722-44-9
Physical Form Neat
Linear Formula C18H31 NO4
MDL Number MFCD00865795
Synonym 1-{4-{[2-(1-Methylethoxy)ethoxy]methyl}phenoxy}-3-[(1-methylethyl)amino]-2-propanol
Type Analytical Standard
Quantity 25 mg
Formula Weight 325.44
Percent Purity ≥98% (HPLC)
Grade Analytical Standard