missing translation for 'onlineSavingsMsg'
Learn More

Bismuthiol II Hydrate 98.0+%, TCI America™

Catalog No. p-7118863
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B11245G 5 g
B112425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B11245G Supplier TCI America Supplier No. B11245G
Only null left

Chemical Identifiers

CAS 6336-51-2
Molecular Formula C8H5KN2S3
Molecular Weight (g/mol) 264.42
MDL Number MFCD00014092
InChI Key QSKDCVGFAPHSHX-UHFFFAOYSA-M
Synonym 1,3,4-thiadiazolidine-2,5-dithione, 3-phenyl-, potassium salt, 3-phenyl-1,3,4-thiadiazolidine-2,5-dithione potassium, potassium 5-mercapto-3-phenyl-1,3,4-thiadiazole-2-thione, 3-phenyl-5-sulfanyl-1,3,4-thiadiazole-2-thione potassium, 5-mercapto-3-phenyl-1,3,4-thiadiazole-2 3h thione potassium salt
PubChem CID 2775614
IUPAC Name potassium;4-phenyl-5-sulfanylidene-1,3,4-thiadiazole-2-thiolate
SMILES C1=CC=C(C=C1)N2C(=S)SC(=N2)[S-].[K+]

Specifications

Quantity 5 g
Formula Weight 264.42
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Bismuthiol II Hydrate
RTECSNumber XI4815000
TSCA No
Recommended Storage Refrigerator