missing translation for 'onlineSavingsMsg'
Learn More

Bismuth Trioxide, 95%, Spectrum™ Chemical

Catalog No. 18609130
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18609130 500 g Amber Glass Bottle
18609128 2.5 kg Amber Glass Bottle
18609129 12 kg Poly Pail
18609131 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18609130 Supplier Spectrum Chemical Mfg Cor Supplier No. B1120500GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 1304-76-3

Specifications

CAS Max % 1
Quantity 500 g
Percent Purity 95%
Grade Reagent
Loss on Drying 0.001
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.