missing translation for 'onlineSavingsMsg'
Learn More

Bismuth Subsalicylate, Purified, 56.5-60.5%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor BI152500GM

 View more versions of this product

Catalog No. 18609119


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

14882-18-9
500 g
Purified
Amber Glass Bottle
1
56.5 to 60.5%
0.01

Chemical Identifiers

14882-18-9
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.