missing translation for 'onlineSavingsMsg'
Learn More

Bismuth Subsalicylate, Purified, 56.5-60.5%, Spectrum™ Chemical

Catalog No. p-7165245
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18609118 125 g Amber Glass Bottle
18609119 500 g Amber Glass Bottle
18609120 2.5 kg Poly Pail
18609121 12 kg Fiber Drum
18609122 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18609118 Supplier Spectrum Chemical Mfg Cor Supplier No. BI152125GM
Only null left

Chemical Identifiers

CAS 14882-18-9

Specifications

CAS Max % 1
Quantity 125 g
Percent Purity 56.5 to 60.5%
Grade Purified
Loss on Drying 0.01
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.