missing translation for 'onlineSavingsMsg'
Learn More

Bismuth Subgallate, Powder, Purified, Approx. 55%, Spectrum™ Chemical

$568.20 - $30651.87

Chemical Identifiers


Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18609111
View Documents
Spectrum Chemical Mfg Cor
BI145125GM
125 g Amber Glass Bottle
Each for $568.20
Only null left
 
18609112
View Documents
Spectrum Chemical Mfg Cor
BI145500GM
500 g Amber Glass Bottle
Each for $1,443.63
Only null left
 
18609113
View Documents
Spectrum Chemical Mfg Cor
BI14525KG
2.5 kg Poly Pail
Each for $4,446.63
Only null left
 
18609114
View Documents
Spectrum Chemical Mfg Cor
BI14550KGBL
50 kg Fiber Drum N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

0.0

Specifications

Amber Glass Bottle
125 g
∼55%
Purified
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.