missing translation for 'onlineSavingsMsg'
Learn More

Bismuth Subgallate, Powder, Purified, Approx. 55%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor BI14525KG

 View more versions of this product

Catalog No. 18609113


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

99-26-3
2.5 kg
Purified
Poly Pail
∼55%

Chemical Identifiers

99-26-3
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.