missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bismuth Nitrate, Pentahydrate, Crystal, ACS, 98%, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor B111525KG
Specifications
| 10035-06-0 | |
| 2.5 kg | |
| FBXVOTBTGXARNA-UHFFFAOYSA-N | |
| bismuth(3+) pentahydrate trinitrate | |
| 98% | |
| Amber Glass Bottle |
| 1 | |
| BiH10N3O14 | |
| O.O.O.O.O.[Bi+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
| 485.07 | |
| ACS |
Chemical Identifiers
| 10035-06-0 | |
| 485.07 | |
| bismuth(3+) pentahydrate trinitrate |
| BiH10N3O14 | |
| FBXVOTBTGXARNA-UHFFFAOYSA-N | |
| O.O.O.O.O.[Bi+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.