missing translation for 'onlineSavingsMsg'
Learn More

Bismuth Nitrate, Pentahydrate, Crystal, ACS, 98%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor B1115500GM

 View more versions of this product

Catalog No. 18609102


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

10035-06-0
500 g
FBXVOTBTGXARNA-UHFFFAOYSA-N
bismuth(3+) pentahydrate trinitrate
98%
Amber Glass Bottle
1
BiH10N3O14
O.O.O.O.O.[Bi+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
485.07
ACS

Chemical Identifiers

10035-06-0
485.07
bismuth(3+) pentahydrate trinitrate
BiH10N3O14
FBXVOTBTGXARNA-UHFFFAOYSA-N
O.O.O.O.O.[Bi+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.