missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bismuth(III) sulfide, 99%, pure
CAS: 1345-07-9 | Bi2S3 | 514.14 g/mol
$252.74 - $252.74
Chemical Identifiers
| CAS | 1345-07-9 |
|---|---|
| Molecular Formula | Bi2S3 |
| Molecular Weight (g/mol) | 514.14 |
| MDL Number | MFCD00014204 |
| InChI Key | YNRGZHRFBQOYPP-UHFFFAOYSA-N |
| IUPAC Name | dibismuth(3+) trisulfanediide |
| SMILES | [S--].[S--].[S--].[Bi+3].[Bi+3] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC453991000
|
Thermo Scientific Chemicals
453991000 |
100 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Chemical Identifiers
| 1345-07-9 | |
| 514.14 | |
| YNRGZHRFBQOYPP-UHFFFAOYSA-N | |
| [S--].[S--].[S--].[Bi+3].[Bi+3] |
| Bi2S3 | |
| MFCD00014204 | |
| dibismuth(3+) trisulfanediide |
Specifications
| 1345-07-9 | |
| 80.1 to 82.5% (Bi) (Complexometry) | |
| Bi2S3 | |
| 15,1287 | |
| [S--].[S--].[S--].[Bi+3].[Bi+3] | |
| 514.14 | |
| 99% | |
| Bismuth(III) sulfide |
| 100 g | |
| Bi2S3 | |
| MFCD00014204 | |
| YNRGZHRFBQOYPP-UHFFFAOYSA-N | |
| dibismuth(3+) trisulfanediide | |
| 514.16 | |
| Pure |
Safety and Handling
EINECSNumber : 215-716-
RUO – Research Use Only