CAS RN 1345-07-9
CAS RN 1345-07-9
IUPAC Name:
dibismuth(3+) trisulfanediide
Synonyms:
bismuth sulfide
Molecular Weight (g/mol):
514.14
Molecular Formula:
Bi2S3
InChi Key:
YNRGZHRFBQOYPP-UHFFFAOYSA-N
SMILES:
[S--].[S--].[S--].[Bi+3].[Bi+3]
1
–
2
of
2
results
1
Bismuth(III) sulfide, 99.9% (metals basis)
CAS: 1345-07-9 Molecular Formula: Bi2S3 Molecular Weight (g/mol): 514.14 MDL Number: MFCD00014204 InChI Key: YNRGZHRFBQOYPP-UHFFFAOYSA-N IUPAC Name: dibismuth(3+) trisulfanediide SMILES: [S--].[S--].[S--].[Bi+3].[Bi+3]
2
Bismuth(III) sulfide, 99%, pure
CAS: 1345-07-9 Molecular Formula: Bi2S3 Molecular Weight (g/mol): 514.14 MDL Number: MFCD00014204 InChI Key: YNRGZHRFBQOYPP-UHFFFAOYSA-N IUPAC Name: dibismuth(3+) trisulfanediide SMILES: [S--].[S--].[S--].[Bi+3].[Bi+3]