missing translation for 'onlineSavingsMsg'
Learn More

Bismuth(III) sulfide, 99%, pure

Catalog No. AC453991000
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC453991000 100 g
AC453995000 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC453991000 Supplier Thermo Scientific Chemicals Supplier No. 453991000
Only null left

CAS: 1345-07-9 | Bi2S3 | 514.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Chemical Identifiers

CAS 1345-07-9
Molecular Formula Bi2S3
Molecular Weight (g/mol) 514.14
MDL Number MFCD00014204
InChI Key YNRGZHRFBQOYPP-UHFFFAOYSA-N
IUPAC Name dibismuth(3+) trisulfanediide
SMILES [S--].[S--].[S--].[Bi+3].[Bi+3]

Specifications

Quantity 100 g
Assay Percent Range 80.1 to 82.5% (Bi) (Complexometry)
Linear Formula Bi2S3
Merck Index 15,1287
Formula Weight 514.16
Percent Purity 99%
Grade Pure
Chemical Name or Material Bismuth(III) sulfide
EINECSNumber 215-716-

RUO – Research Use Only