missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bismuth(III) sulfate, 99%, pure, Thermo Scientific Chemicals
$264.38 - $264.38
Chemical Identifiers
| CAS | 7787-68-0 |
|---|---|
| Molecular Formula | Bi2O12S3 |
| Molecular Weight (g/mol) | 706.13 |
| MDL Number | MFCD00036140 |
| InChI Key | BEQZMQXCOWIHRY-UHFFFAOYSA-H |
| Synonym | bismuth sulfate, dibismuth trisulfate, bismuth trisulfate, sulfuric acid, bismuth 3+ salt 3:2, bismuth sulphate, bismuth sesquisulfate, bismuth sulfate mi, dibismuth tris sulphate, bismuth iii sulfate, 2bi.3so4 |
| PubChem CID | 198144 |
| IUPAC Name | dibismuth;trisulfate |
| SMILES | [Bi+3].[Bi+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC453851000
|
Thermo Scientific Chemicals
453851000 |
100 g |
Each for $264.38
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 7787-68-0 | |
| 706.13 | |
| BEQZMQXCOWIHRY-UHFFFAOYSA-H | |
| 198144 | |
| [Bi+3].[Bi+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O |
| Bi2O12S3 | |
| MFCD00036140 | |
| bismuth sulfate, dibismuth trisulfate, bismuth trisulfate, sulfuric acid, bismuth 3+ salt 3:2, bismuth sulphate, bismuth sesquisulfate, bismuth sulfate mi, dibismuth tris sulphate, bismuth iii sulfate, 2bi.3so4 | |
| dibismuth;trisulfate |
Specifications
| 405.0°C | |
| 100 g | |
| Bi2O12S3 | |
| MFCD00036140 | |
| bismuth sulfate, dibismuth trisulfate, bismuth trisulfate, sulfuric acid, bismuth 3+ salt 3:2, bismuth sulphate, bismuth sesquisulfate, bismuth sulfate mi, dibismuth tris sulphate, bismuth iii sulfate, 2bi.3so4 | |
| [Bi+3].[Bi+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O | |
| 706.13 | |
| 706.13 | |
| Pure |
| 7787-68-0 | |
| 98.5% min | |
| Bi2(SO4)3 | |
| 15,1286 | |
| BEQZMQXCOWIHRY-UHFFFAOYSA-H | |
| dibismuth;trisulfate | |
| 198144 | |
| 99% | |
| Bismuth(III) sulfate |
Safety and Handling
EINECSNumber : 232-129-5
RUO – Research Use Only