accessibility menu, dialog, popup

UserName

BiBiOSOOOOSOOOOSOOO

CAS RN 7787-68-0

IUPAC Name: dibismuth(3+) trisulfate
Synonyms: dibismuth(3+) trisulfate
Molecular Weight (g/mol): 706.13
Molecular Formula: Bi2O12S3
InChi Key: BEQZMQXCOWIHRY-UHFFFAOYSA-H
SMILES: [Bi+3].[Bi+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)