missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bismuth(Iii) Subnitrate, Honeywell Fluka™
Puriss., meets analytical specification of DAB, USP, 71-74% Bi basis (calc. on dried substance)
$267.53 - $893.32
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
| 1304-85-4 | |
| MFCD00064844 |
Specifications
| 1304-85-4 | |
| Bi5O(OH)9(NO3)4 | |
| 1477 | |
| puriss. | |
| Plastic Bottle |
| 250 g | |
| MFCD00064844 | |
| 1461.99g/mol | |
| 71 to 74% Bi basis (calc. on dried substance) | |
| Bismuth(Iii) Subnitrate, Meets analytical specification of USP |
Safety and Handling
P220-P261-P305 + P351 + P338
H272-H315-H319-H335
EINECSNumber : 215-136-8