missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bismuth(Iii) Subnitrate, Solstice
Description
- Leading the Industry For Over 65 Years
- Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Quantity | 250 g |
| Linear Formula | Bi5O(OH)9(NO3)4 |
| UN Number | 1477 |
| Formula Weight | 1461.99g/mol |
| Grade | puriss. |
| Concentration or Composition (by Analyte or Components) | 71 to 74% Bi basis (calc. on dried substance) |
| Packaging | Plastic Bottle |
| Chemical Name or Material | Bismuth(Iii) Subnitrate, Meets analytical specification of USP |
Safety and Handling
| Hazard Statement | P220-P261-P305 + P351 + P338 |
| Precautionary Statement | H272-H315-H319-H335 |
| EINECSNumber | 215-136-8 |