missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bismuth(III) oxyiodide, 98%, pure, Thermo Scientific™
Chemical Identifiers
| CAS | 7787-63-5 |
|---|---|
| Molecular Formula | BiIO |
| Molecular Weight (g/mol) | 351.88 |
| InChI Key | FCBARUADRQNVFT-UHFFFAOYSA-N |
| Synonym | bismuth iodide oxide, biio, bismuth iii oxyiodide, bismuth oxyiodide, bismuth oxyiodide, bioi |
| PubChem CID | 6367235 |
| IUPAC Name | oxobismuth;hydroiodide |
| SMILES | O=[Bi].I |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AC453830100
|
N/A | 10g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 7787-63-5 | |
| 351.88 | |
| bismuth iodide oxide, biio, bismuth iii oxyiodide, bismuth oxyiodide, bismuth oxyiodide, bioi | |
| oxobismuth;hydroiodide |
| BiIO | |
| FCBARUADRQNVFT-UHFFFAOYSA-N | |
| 6367235 | |
| O=[Bi].I |
Specifications
| 7787-63-5 | |
| BiIO | |
| 15,127 | |
| Solubility in water: insoluble. | |
| O=[Bi].I | |
| 351.88 | |
| 351.88 | |
| Pure |
| 97.5% min | |
| IBi=O | |
| bismuth iodide oxide, biio, bismuth iii oxyiodide, bismuth oxyiodide, bismuth oxyiodide, bioi | |
| FCBARUADRQNVFT-UHFFFAOYSA-N | |
| oxobismuth;hydroiodide | |
| 6367235 | |
| 98% | |
| Bismuth(III) oxyiodide |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse
GHS Signal Word: Warning
EINECSNumber : 232-126-9
RUO – Research Use Only