missing translation for 'onlineSavingsMsg'
Learn More

Bismuth(III) oxyiodide, 98%, pure, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 453830100

Quantity 10g

 View more versions of this product

Catalog No. AC453830100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

7787-63-5
351.88
bismuth iodide oxide, biio, bismuth iii oxyiodide, bismuth oxyiodide, bismuth oxyiodide, bioi
oxobismuth;hydroiodide
BiIO
FCBARUADRQNVFT-UHFFFAOYSA-N
6367235
O=[Bi].I

Specifications

Bismuth(III) oxyiodide
10g
BiIO
15,127
Solubility in water: insoluble.
O=[Bi].I
351.88
351.88
Pure
7787-63-5
97.5% min
IBi=O
bismuth iodide oxide, biio, bismuth iii oxyiodide, bismuth oxyiodide, bismuth oxyiodide, bioi
FCBARUADRQNVFT-UHFFFAOYSA-N
oxobismuth;hydroiodide
6367235
98%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse

GHS Signal Word: Warning

EINECSNumber : 232-126-9

SDS
Documents

Documents

Promotions

Promotions