missing translation for 'onlineSavingsMsg'
Learn More

Bismuth(III) oxide, typically 99.99% (metals basis)

CAS: 1304-76-3 | Bi2O3 | 465.96 g/mol

$148.81 - $561.37

Chemical Identifiers

CAS 1304-76-3
Molecular Formula Bi2O3
Molecular Weight (g/mol) 465.96
MDL Number MFCD00003462
InChI Key TYIXMATWDRGMPF-UHFFFAOYSA-N
IUPAC Name dibismuth(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Bi+3].[Bi+3]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1713218
View Documents
Thermo Scientific Chemicals
01713218
50 g
Each for $148.81
Only null left
 
AA1713230
View Documents
Thermo Scientific Chemicals
01713230
250 g
Each for $561.37
Only null left
 
Description

Description

Used as an adhesive and sealant chemical, flame retardant, pigment, propellant and blowing agent. It is also used as a solvent. Other uses are in batteries, electrical and electronic products, explosive materials, ink, toner, and colorant products, paints and coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1304-76-3
465.96
TYIXMATWDRGMPF-UHFFFAOYSA-N
[O--].[O--].[O--].[Bi+3].[Bi+3]
Bi2O3
MFCD00003462
dibismuth(3+) trioxidandiide

Specifications

ca 1890°C
1304-76-3
50 g
Bi2O3
Insoluble in water. Soluble in acids.
[O--].[O--].[O--].[Bi+3].[Bi+3]
465.96
8.9
817°C
Powder
99.99% (metals basis)
MFCD00003462
TYIXMATWDRGMPF-UHFFFAOYSA-N
dibismuth(3+) trioxidandiide
465.96
Bismuth(III) oxide
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 215-134-7

RTECSNumber : EB2984460

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only