missing translation for 'onlineSavingsMsg'
Learn More

Bismuth(III) oxide, Puratronic™, 99.999% (metals basis)

Catalog No. AA4286318
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4286318 50 g
AA4286330 250 g
AA42863A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4286318 Supplier Thermo Scientific Chemicals Supplier No. 04286318

CAS: 1304-76-3 | Bi2O3 | 465.96 g/mol

Bismuth(III) oxide is used in solid-oxide fuel cells. It is also used in optical glasses, to replace lead oxide in whitewares, luxes, varistor formulations and ceramic capacitor formulations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1304-76-3
Molecular Formula Bi2O3
Molecular Weight (g/mol) 465.96
MDL Number MFCD00003462
InChI Key TYIXMATWDRGMPF-UHFFFAOYSA-N
IUPAC Name dibismuth(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Bi+3].[Bi+3]

Specifications

Boiling Point ∼1890°C
Melting Point 817°C
Color Yellow
Physical Form Powder
Quantity 50 g
Merck Index 14,1273
Solubility Information Insoluble in water. Soluble in acids.
Formula Weight 465.96
Percent Purity 99.999%
Grade Puratronic
Density 8.9 g/mL
Assay (metals basis)
Chemical Name or Material Bismuth (III) oxide
Show More Show Less
EINECSNumber 215-134-7
RTECSNumber EB2984460
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only