missing translation for 'onlineSavingsMsg'
Learn More

Bismuth(III) Oxide, Extra Pure, MilliporeSigma™

Catalog No. M1018629025
Change view
Click to view available options
Quantity:
25 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1018629025 25 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1018629025 Supplier Technidata Supplier No. 1.01862.9025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High-purity bismuth(III) oxide suitable for general laboratory applications.

Chemical Identifiers

CAS 1304-76-3
Molecular Formula Bi2O3
Molecular Weight (g/mol) 465.96
MDL Number MFCD00003462
InChI Key TYIXMATWDRGMPF-UHFFFAOYSA-N
IUPAC Name dibismuth(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Bi+3].[Bi+3]

Specifications

Physical Form Powder
Quantity 25 kg
Odor Odorless
Packaging Fibre carton
Appearance Yellow powder
Formula Weight 465.95
Percent Purity ≥99.0%
Grade Extra Pure
Chemical Name or Material Bismuth(III) oxide