Learn More
Bismuth(III) nitrate pentahydrate, 98%, pure, Thermo Scientific™
$247.49 - $247.49
Chemical Identifiers
| CAS | 10035-06-0 |
|---|---|
| Molecular Formula | BiH10N3O14 |
| Molecular Weight (g/mol) | 485.07 |
| MDL Number | MFCD00149157 |
| InChI Key | FBXVOTBTGXARNA-UHFFFAOYSA-N |
| Synonym | bismuth nitrate pentahydrate, bismuth trinitrate pentahydrate, bismuth nitrate, hydrate 1:5, bismuth iii nitrate pentahydrate, unii-fo2icy167b, fo2icy167b, nitric acid, bismuth 3+ salt, pentahydrate, acmc-20alle, bismuthnitratepentahydrate |
| PubChem CID | 160911 |
| IUPAC Name | bismuth;trinitrate;pentahydrate |
| SMILES | O.O.O.O.O.[Bi+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC454110010
|
Thermo Scientific Chemicals
454110010 |
1kg |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 10035-06-0 | |
| 485.07 | |
| FBXVOTBTGXARNA-UHFFFAOYSA-N | |
| 160911 | |
| O.O.O.O.O.[Bi+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O |
| BiH10N3O14 | |
| MFCD00149157 | |
| bismuth nitrate pentahydrate, bismuth trinitrate pentahydrate, bismuth nitrate, hydrate 1:5, bismuth iii nitrate pentahydrate, unii-fo2icy167b, fo2icy167b, nitric acid, bismuth 3+ salt, pentahydrate, acmc-20alle, bismuthnitratepentahydrate | |
| bismuth;trinitrate;pentahydrate |
Specifications
| 10035-06-0 | |
| 97.5% min. (Complexometry) | |
| Bi(NO3)3·5H2O | |
| 15,1271 | |
| Solubility in water: may decompose. | |
| O.O.O.O.O.[Bi+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
| 485.07 | |
| 485.07 | |
| Pure |
| 1kg | |
| BiH10N3O14 | |
| MFCD00149157 | |
| bismuth nitrate pentahydrate, bismuth trinitrate pentahydrate, bismuth nitrate, hydrate 1:5, bismuth iii nitrate pentahydrate, unii-fo2icy167b, fo2icy167b, nitric acid, bismuth 3+ salt, pentahydrate, acmc-20alle, bismuthnitratepentahydrate | |
| FBXVOTBTGXARNA-UHFFFAOYSA-N | |
| bismuth;trinitrate;pentahydrate | |
| 160911 | |
| 98% | |
| Bismuth(III) nitrate pentahydrate |
Safety and Handling
GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. May intensify fire; oxidizer.
GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Avoid breathing dust/fume/gas/mist/vapors/spray. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
GHS Signal Word: Danger
RUO â Research Use Only