missing translation for 'onlineSavingsMsg'
Learn More

Bismuth(III) acetate, 99%

CAS: 22306-37-2 | C6H9BiO6 | 386.112 g/mol

$250.67 - $827.93

Chemical Identifiers

CAS 22306-37-2
Molecular Formula C6H9BiO6
Molecular Weight (g/mol) 386.112
MDL Number MFCD00015629
InChI Key WKLWZEWIYUTZNJ-UHFFFAOYSA-K
Synonym bismuth acetate, bismuth triacetate, unii-8aja86y692, acetic acid, bismuth 3+ salt, acmc-20allk, bismuth iii ; acetate, bismuth 3+ triacetate, trisacetic acid bismuth salt, bismuth 3+ ion tris acetate ion, bismuth iii acetate 50g
PubChem CID 31132
IUPAC Name bismuth;triacetate
SMILES CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].[Bi+3]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1757418
View Documents
Thermo Scientific Chemicals
01757418
50 g
Each for $250.67
Only null left
 
AA1757430
View Documents
Thermo Scientific Chemicals
01757430
250 g
Each for $827.93
Only null left
 
Description

Description

Bismuth(III) acetate is used in biology, printing, in photographic films, textiles, and also in proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

22306-37-2
386.112
WKLWZEWIYUTZNJ-UHFFFAOYSA-K
31132
CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].[Bi+3]
C6H9BiO6
MFCD00015629
bismuth acetate, bismuth triacetate, unii-8aja86y692, acetic acid, bismuth 3+ salt, acmc-20allk, bismuth iii ; acetate, bismuth 3+ triacetate, trisacetic acid bismuth salt, bismuth 3+ ion tris acetate ion, bismuth iii acetate 50g
bismuth;triacetate

Specifications

(decomposition)
22306-37-2
50 g
Bi(OOCCH3)3
bismuth acetate, bismuth triacetate, unii-8aja86y692, acetic acid, bismuth 3+ salt, acmc-20allk, bismuth iii ; acetate, bismuth 3+ triacetate, trisacetic acid bismuth salt, bismuth 3+ ion tris acetate ion, bismuth iii acetate 50g
WKLWZEWIYUTZNJ-UHFFFAOYSA-K
bismuth;triacetate
31132
99%
Bismuth(III) acetate
White
Crystalline
C6H9BiO6
MFCD00015629
Soluble in water.
CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].[Bi+3]
386.112
386.12
Hygroscopic
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 244-904-5

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only