missing translation for 'onlineSavingsMsg'
Learn More

Bismuth hydroxide, tech.

Catalog No. AA4039730
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4039730 250 g
AA40397A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4039730 Supplier Thermo Scientific Chemicals Supplier No. 04039730

Hydrolysis of ribonucleic acid, absorbent | CAS: 10361-43-0 | BiH3O3 | 260.00 g/mol

It is used for the hydrolysis of ribonucleic acid and as absorbent for rutin and quercetin. It is a component used in milk of bismuth is used in gastrointestinal disorders as a protective agent. It is used for the removal of nitrate and other anions from water by yellow bismuth hydroxide. A bismuth oxide nanoplate is useful for carbon dioxide gas sensor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10361-43-0
Molecular Formula BiH3O3
Molecular Weight (g/mol) 260.00
MDL Number MFCD00046188
InChI Key TZSXPYWRDWEXHG-UHFFFAOYSA-K
IUPAC Name bismuth(3+) trihydroxide
SMILES [OH-].[OH-].[OH-].[Bi+3]

Specifications

Physical Form Amorphous Powder
Quantity 250 g
Solubility Information Insoluble in water; Soluble in acids.
Formula Weight 260
Grade Technical
Odor Odorless
Chemical Name or Material Bismuth hydroxide
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only