CAS RN 10361-43-0
IUPAC Name:
bismuth(3+) trihydroxide
Synonyms:
bismuth hydroxide
Molecular Weight (g/mol):
260
Molecular Formula:
BiH3O3
InChi Key:
TZSXPYWRDWEXHG-UHFFFAOYSA-K
SMILES:
[OH-].[OH-].[OH-].[Bi+3]
1
–
2
of
2
results
1
Bismuth hydroxide, tech.
CAS: 10361-43-0 Molecular Formula: BiH3O3 Molecular Weight (g/mol): 260.00 MDL Number: MFCD00046188 InChI Key: TZSXPYWRDWEXHG-UHFFFAOYSA-K IUPAC Name: bismuth(3+) trihydroxide SMILES: [OH-].[OH-].[OH-].[Bi+3]
2
Promotions Available
Bismuth hydroxide
Bismuth hydroxide
CAS: 10361-43-0 Molecular Formula: BiH3O3 Molecular Weight (g/mol): 260.00 MDL Number: MFCD00046188 InChI Key: TZSXPYWRDWEXHG-UHFFFAOYSA-K IUPAC Name: bismuth(3+) trihydroxide SMILES: [OH-].[OH-].[OH-].[Bi+3]