Learn More
Bismuth antimonide, 99.99%
CAS: 12323-19-2 | Bi2Sb2 | 661.48 g/mol
$111.04 - $492.52
Chemical Identifiers
| CAS | 12323-19-2 |
|---|---|
| Molecular Formula | Bi2Sb2 |
| Molecular Weight (g/mol) | 661.48 |
| MDL Number | MFCD00014196 |
| InChI Key | AEMQIQQWIVNHAU-UHFFFAOYSA-N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1701409
|
Thermo Scientific Chemicals
A1701409 |
10 g |
Each for $111.04
|
|
|||||
|
AAA1701418
|
Thermo Scientific Chemicals
A1701418 |
50 g |
Each for $492.52
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 12323-19-2 | |
| Bi2Sb2 | |
| UN1549 | |
| 661.48 | |
| 99.99% |
| 10 g | |
| MFCD00014196 | |
| AEMQIQQWIVNHAU-UHFFFAOYSA-N | |
| 330.73 | |
| Bismuth antimonide |
Safety and Handling
GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
H302+H332
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ANTIMONY COMPOUNDS, INORGANIC, SOLID, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only