accessibility menu, dialog, popup

UserName

SbSbBiBi

CAS RN 12323-19-2

IUPAC Name: 1,2-distiba-3,4-dibismatricyclo[1.1.0.0²,⁴]butane
Synonyms: 1,2-distiba-3,4-dibismatricyclo[1.1.0.0²,⁴]butane
Molecular Weight (g/mol): 661.48
Molecular Formula: Bi2Sb2
InChi Key: AEMQIQQWIVNHAU-UHFFFAOYSA-N
SMILES: [Sb]12[Sb]3[Bi]1[Bi]23

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results