missing translation for 'onlineSavingsMsg'
Learn More

Bismarck Brown Y

Catalog No. aaa1667422
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1667422 100 g
AAA1667414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1667422 Supplier Thermo Scientific Chemicals Supplier No. A1667422
Only null left

CAS: 10114-58-6 | C18H20Cl2N8 | 419.314 g/mol

Plasma stain for mucins, cartilage and goblet cellsBismarck Brown Y is used for staining mucine, cartilage and goblet cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Plasma stain for mucins, cartilage and goblet cellsBismarck Brown Y is used for staining mucine, cartilage and goblet cells.

Solubility
Soluble in water. Slightly soluble in ethanol and cellosolve. Insoluble in acetone, carbon tetrachloride and benzene.

Notes
Incompatible with strong oxidizing agents and reducing agents.

Chemical Identifiers

CAS 10114-58-6
Molecular Formula C18H20Cl2N8
Molecular Weight (g/mol) 419.314
MDL Number MFCD00012977
InChI Key MCZVRBLCRZWFJH-UHFFFAOYSA-N
Synonym C.I. 21000
PubChem CID 82360
ChEBI CHEBI:53615
IUPAC Name 4-[[3-[(2,4-diaminophenyl)diazenyl]phenyl]diazenyl]benzene-1,3-diamine;dihydrochloride
SMILES C1=CC(=CC(=C1)N=NC2=C(C=C(C=C2)N)N)N=NC3=C(C=C(C=C3)N)N.Cl.Cl

Specifications

Merck Index 14,1253
Solubility Information Soluble in water; Slightly soluble in ethanol and cellosolve; Insoluble in acetone,carbon tetrachloride and benzene.
Formula Weight 419.32
Odor Odorless
Quantity 100 g
Chemical Name or Material Bismarck Brown Y
Hazard Category H228-H302+H312+H332
Precautionary Statement P210-P240-P241-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P370+P378q-P501c
EINECSNumber 233-314-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only