missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bisdemethoxycurcumin 98.0+%, TCI America™
Supplier: TCI America B334725G
Specifications
| Bisdemethoxycurcumin | |
| 224°C | |
| MFCD03419284 | |
| 1,7-Bis(4-hydroxyphenyl)-1,6-heptadiene-3,5-dione, Didemethoxycurcumin | |
| C1=CC(=CC=C1C=CC(=O)CC(=O)C=CC2=CC=C(C=C2)O)O | |
| 308.333 | |
| CHEBI:71045 | |
| ≥98.0% (HPLC,T) |
| 24939-16-0 | |
| C19H16O4 | |
| 25 g | |
| PREBVFJICNPEKM-YDWXAUTNSA-N | |
| (1E,6E)-1,7-bis(4-hydroxyphenyl)hepta-1,6-diene-3,5-dione | |
| 5315472 | |
| 308.33 | |
| Crystalline Powder |
Chemical Identifiers
| 24939-16-0 | |
| 308.333 | |
| PREBVFJICNPEKM-YDWXAUTNSA-N | |
| 5315472 | |
| (1E,6E)-1,7-bis(4-hydroxyphenyl)hepta-1,6-diene-3,5-dione |
| C19H16O4 | |
| MFCD03419284 | |
| 1,7-Bis(4-hydroxyphenyl)-1,6-heptadiene-3,5-dione, Didemethoxycurcumin | |
| CHEBI:71045 | |
| C1=CC(=CC=C1C=CC(=O)CC(=O)C=CC2=CC=C(C=C2)O)O |
Safety and Handling
TSCA : No