missing translation for 'onlineSavingsMsg'
Learn More

Bisdemethoxycurcumin 98.0+%, TCI America™

Catalog No. p-7122892
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B33475G 5 g
B334725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B33475G Supplier TCI America Supplier No. B33475G
Only null left

Chemical Identifiers

CAS 24939-16-0
Molecular Formula C19H16O4
Molecular Weight (g/mol) 308.333
MDL Number MFCD03419284
InChI Key PREBVFJICNPEKM-YDWXAUTNSA-N
Synonym 1,7-Bis(4-hydroxyphenyl)-1,6-heptadiene-3,5-dione, Didemethoxycurcumin
PubChem CID 5315472
ChEBI CHEBI:71045
IUPAC Name (1E,6E)-1,7-bis(4-hydroxyphenyl)hepta-1,6-diene-3,5-dione
SMILES C1=CC(=CC=C1C=CC(=O)CC(=O)C=CC2=CC=C(C=C2)O)O

Specifications

Melting Point 224°C
Quantity 5 g
Formula Weight 308.33
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Bisdemethoxycurcumin