missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Bisbenzimide H 33258 Fluorochrome, Trihydrochloride Calbiochem™,

Membrane-permeable, adenine-thymine-specific fluorescent stain

$181.00 - $1230.00

Chemical Identifiers

CAS 23491-45-4
Molecular Formula C25H37Cl3N6O6
Molecular Weight (g/mol) 623.957
InChI Key URHMSNPWJDDZSH-UHFFFAOYSA-N
Synonym bisbenzimide h 33258 fluorochrome trihydrochloride
PubChem CID 18386398
IUPAC Name 4-[5-[6-(4-methylpiperazin-1-yl)-1H-benzimidazol-2-yl]-1,3-dihydrobenzimidazol-2-ylidene]cyclohexa-2,5-dien-1-one;pentahydrate;trihydrochloride
SMILES CN1CCN(CC1)C2=CC3=C(C=C2)N=C(N3)C4=CC5=C(C=C4)NC(=C6C=CC(=O)C=C6)N5.O.O.O.O.O.Cl.Cl.Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
382061100MG
View Documents
MilliporeSigma™
382061100MG
100 mg
Each for $181.00
Only null left
 
382061250MG
View Documents
MilliporeSigma™
382061250MG
250 mg
Each for $447.00
Only null left
 
3820611GM
View Documents
MilliporeSigma™
3820611GM
1 g
Each for $1,230.00
Only null left
 
Description

Description

Specifications

Chemical Identifiers

23491-45-4
623.957
bisbenzimide h 33258 fluorochrome trihydrochloride
4-[5-[6-(4-methylpiperazin-1-yl)-1H-benzimidazol-2-yl]-1,3-dihydrobenzimidazol-2-ylidene]cyclohexa-2,5-dien-1-one;pentahydrate;trihydrochloride
C25H37Cl3N6O6
URHMSNPWJDDZSH-UHFFFAOYSA-N
18386398
CN1CCN(CC1)C2=CC3=C(C=C2)N=C(N3)C4=CC5=C(C=C4)NC(=C6C=CC(=O)C=C6)N5.O.O.O.O.O.Cl.Cl.Cl

Specifications

23491-45-4
C25H37Cl3N6O6
bisbenzimide h 33258 fluorochrome trihydrochloride
URHMSNPWJDDZSH-UHFFFAOYSA-N
4-[5-[6-(4-methylpiperazin-1-yl)-1H-benzimidazol-2-yl]-1,3-dihydrobenzimidazol-2-ylidene]cyclohexa-2,5-dien-1-one;pentahydrate;trihydrochloride
18386398
≥98%
Solid
Yellow
100 mg
Water: soluble; Precipitates in phosphate buffered solutions
CN1CCN(CC1)C2=CC3=C(C=C2)N=C(N3)C4=CC5=C(C=C4)NC(=C6C=CC(=O)C=C6)N5.O.O.O.O.O.Cl.Cl.Cl
623.957
533.9
HPLC
Bisbenzimide H 33258 Fluorochrome
Videos
Safety and Handling

Safety and Handling

Recommended Storage : 2° to 8°C (35.6° to 46.4°F)

SDS
missing translation for 'documents'

missing translation for 'documents'