missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Bisbenzimide H 33258 Fluorochrome, Trihydrochloride Calbiochem™,
Membrane-permeable, adenine-thymine-specific fluorescent stain
Supplier: MilliporeSigma™ 3820611GM
Specifications
| Bisbenzimide H 33258 Fluorochrome | |
| Yellow | |
| 1 g | |
| Water: soluble; Precipitates in phosphate buffered solutions | |
| CN1CCN(CC1)C2=CC3=C(C=C2)N=C(N3)C4=CC5=C(C=C4)NC(=C6C=CC(=O)C=C6)N5.O.O.O.O.O.Cl.Cl.Cl | |
| 623.957 | |
| 533.9 | |
| HPLC |
| 23491-45-4 | |
| C25H37Cl3N6O6 | |
| bisbenzimide h 33258 fluorochrome trihydrochloride | |
| URHMSNPWJDDZSH-UHFFFAOYSA-N | |
| 4-[5-[6-(4-methylpiperazin-1-yl)-1H-benzimidazol-2-yl]-1,3-dihydrobenzimidazol-2-ylidene]cyclohexa-2,5-dien-1-one;pentahydrate;trihydrochloride | |
| 18386398 | |
| ≥98% | |
| Solid |
Chemical Identifiers
| 23491-45-4 | |
| 623.957 | |
| bisbenzimide h 33258 fluorochrome trihydrochloride | |
| 4-[5-[6-(4-methylpiperazin-1-yl)-1H-benzimidazol-2-yl]-1,3-dihydrobenzimidazol-2-ylidene]cyclohexa-2,5-dien-1-one;pentahydrate;trihydrochloride |
| C25H37Cl3N6O6 | |
| URHMSNPWJDDZSH-UHFFFAOYSA-N | |
| 18386398 | |
| CN1CCN(CC1)C2=CC3=C(C=C2)N=C(N3)C4=CC5=C(C=C4)NC(=C6C=CC(=O)C=C6)N5.O.O.O.O.O.Cl.Cl.Cl |
Safety and Handling
Recommended Storage : 2° to 8°C (35.6° to 46.4°F)