missing translation for 'onlineSavingsMsg'
Learn More

Bisacodyl 98.0+%, TCI America™

Catalog No. B50665G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B50665G 5 g
B506625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B50665G Supplier TCI America Supplier No. B50665G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 603-50-9
Molecular Formula C22H19NO4
Molecular Weight (g/mol) 361.40
MDL Number MFCD00038039
InChI Key KHOITXIGCFIULA-UHFFFAOYSA-N
Synonym bisacodyl, dulcolax, bicol, brocalax, durolax, dulcolan, endokolat, fenilaxan, hillcolax, sanvacual
PubChem CID 2391
IUPAC Name 4-{[4-(acetyloxy)phenyl](pyridin-2-yl)methyl}phenyl acetate
SMILES CC(=O)OC1=CC=C(C=C1)C(C1=CC=C(OC(C)=O)C=C1)C1=CC=CC=N1

Specifications

Melting Point 134°C
Color White
Quantity 5 g
Formula Weight 361.40
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Bisacodyl
EINECSNumber (9)-2202
RTECSNumber SM8750000
TSCA Yes