missing translation for 'onlineSavingsMsg'
Learn More

Bisacodyl, MilliporeSigma™ Supelco™

Catalog No. 111014857
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014857 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014857 Supplier Merck Emd Millipore Supplier No. B139010G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Bisacodyl is a diphenylmethane derivative, mainly used as a stimulant laxative for treating constipation.

Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 603-50-9
Physical Form Neat
Linear Formula C22H19 NO4
MDL Number MFCD00038039
Type Analytical Standard
Industry Type Drug Analysis
Quantity 10 g
Formula Weight 361.39
Grade Analytical Standard