missing translation for 'onlineSavingsMsg'
Learn More

Bisacodyl, 98+%

Catalog No. p-7042289
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6199709 10 g
AAJ6199714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6199709 Supplier Thermo Scientific Chemicals Supplier No. J6199709
Only null left

CAS: 603-50-9 | C22H19NO4 | 361.40 g/mol

Bisacodyl is used in the inhibition of (Na+K)ATPase, the increase of mucosal PGE2, and the activation of adenyl cyclase in rat intestine studies. It is used as a laxative drug. It is typically prescribed for relief of constipation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in alcohol (slightly), ether (slightly), chloroform, and acetone. Insoluble in water.

Notes
Protect from heat. Incompatible with heat and oxidizing agents. Store at 4°C.

Chemical Identifiers

CAS 603-50-9
Molecular Formula C22H19NO4
Molecular Weight (g/mol) 361.40
MDL Number MFCD00038039
InChI Key KHOITXIGCFIULA-UHFFFAOYSA-N
Synonym bisacodyl, dulcolax, bicol, brocalax, durolax, dulcolan, endokolat, fenilaxan, hillcolax, sanvacual
PubChem CID 2391
IUPAC Name 4-{[4-(acetyloxy)phenyl](pyridin-2-yl)methyl}phenyl acetate
SMILES CC(=O)OC1=CC=C(C=C1)C(C1=CC=C(OC(C)=O)C=C1)C1=CC=CC=N1

Specifications

Color White
Quantity 10 g
Merck Index 14,1242
Solubility Information Soluble in alcohol (slightly),ether (slightly),chloroform,and acetone. Insoluble in water.
Formula Weight 361.4
Percent Purity ≥98%
Physical Form Crystalline Powder
Chemical Name or Material Bisacodyl
EINECSNumber 210-044-4
RTECSNumber SM8750000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only