missing translation for 'onlineSavingsMsg'
Learn More

Bisabolene, mixture of isomers

Catalog No. p-7024306
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1872409 10 g
AAA1872418 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1872409 Supplier Thermo Scientific Chemicals Supplier No. A1872409
Only null left

CAS: 495-62-5 | C15H24 | 204.357 g/mol

It finds use in artificial oil of bergamot, myrrh and lemon, and is an excellent fixative for neroli bases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds use in artificial oil of bergamot, myrrh and lemon, and is an excellent fixative for neroli bases.

Solubility
Soluble in alcohol, water, (0.008994 mg/L @ 25°C (est)).

Notes
Air Sensitive. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 495-62-5
Molecular Formula C15H24
Molecular Weight (g/mol) 204.357
MDL Number MFCD00129080
InChI Key XBGUIVFBMBVUEG-CCEZHUSRSA-N
Synonym e-gamma-bisabolene, unii-l8m7r52o3b, cyclohexene, 4-1,5-dimethyl-4-hexenylidene-1-methyl-, e, 1e-bisabola-1 10 ,4,7 11-triene 4e-4-1,5-dimethylhex-4-en-1-ylidene-1-methylcyclohexene, gamma-bisabolene, e, gamma-bisabolene, 4e, cyclohexene, 4-1,5-dimethyl-4-hexenylidene-1-methyl-, 4e, .gamma.-bisabolene, e gamma-bisabolene, 1-methyl-4-6-methylhept-5-en-2-ylidene cyclohex-1-ene
PubChem CID 3033866
ChEBI CHEBI:49238
IUPAC Name (4Z)-1-methyl-4-(6-methylhept-5-en-2-ylidene)cyclohexene
SMILES CC1=CCC(=C(C)CCC=C(C)C)CC1

Specifications

Density 0.89
Refractive Index 1.494
Quantity 10 g
Sensitivity Air Sensitive
Solubility Information Soluble in alcohol,water,(0.008994mg/L @ 25°C (est)).
Formula Weight 204.36
Chemical Name or Material Bisabolene, mixture of isomers
Hazard Category H227
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 207-805-8
RTECSNumber GW6060000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only