accessibility menu, dialog, popup

UserName

CAS RN 495-62-5

IUPAC Name: (4Z)-1-methyl-4-(6-methylhept-5-en-2-ylidene)cyclohex-1-ene
Synonyms: (Z)-gamma-bisabolene
Molecular Weight (g/mol): 204.36
Molecular Formula: C15H24
InChi Key: XBGUIVFBMBVUEG-CCEZHUSRSA-N
SMILES: CC(C)=CCC\C(C)=C1\CCC(C)=CC1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results