missing translation for 'onlineSavingsMsg'
Learn More

Bis(triphenylphosphoranylidene)ammonium chloride, 97% (dry wt.), water <3%

CAS: 21050-13-5 | C36H30ClNP2 | 574.04 g/mol

$148.45 - $503.11

Chemical Identifiers

CAS 21050-13-5
Molecular Formula C36H30ClNP2
Molecular Weight (g/mol) 574.04
MDL Number MFCD00151523
InChI Key LVRCYPYRKNAAMX-UHFFFAOYSA-M
Synonym 1,1,1-triphenyl-n-triphenylphosphoranylidene phosphoraniminium chloride, bis triphenylphosphoranylidene ammonium chloride, bis triphenylphosphoranylidene ammonium, triphenyl p,p,p-triphenylphosphine imidato-n phosphorus 1+ tetracarbonylcobaltate 1-, bis triphenylphosphine iminium chloride∼ppncl, ppn chloride, ppn cl, ksc491c1r, parent, triphenyl p,p,p-triphenylphosphine imidato phosphorus 1+ chloride
PubChem CID 3036656
IUPAC Name triphenyl-[(triphenyl-$l^{5}-phosphanylidene)amino]phosphanium;chloride
SMILES [Cl-].C1=CC=C(C=C1)P(=[N+]=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1433906
View Documents
Thermo Scientific Chemicals
L1433906
5 g N/A
Only null left
 
AAL1433914
View Documents
Thermo Scientific Chemicals
L1433914
25 g N/A
Only null left
 
Description

Description

Co-catalyst used with a chiral Cobalt-salen complex for the asymmetric addition of carbon dioxide to propylene oxide. Also used in the preparation of 5-Hydroxy-clethodim Sulfoxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Co-catalyst used with a chiral Cobalt-salen complex for the asymmetric addition of carbon dioxide to propylene oxide. Also used in the preparation of 5-Hydroxy-clethodim Sulfoxide.

Solubility
Moderately soluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Chemical Identifiers

21050-13-5
574.04
LVRCYPYRKNAAMX-UHFFFAOYSA-M
3036656
[Cl-].C1=CC=C(C=C1)P(=[N+]=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
C36H30ClNP2
MFCD00151523
1,1,1-triphenyl-n-triphenylphosphoranylidene phosphoraniminium chloride, bis triphenylphosphoranylidene ammonium chloride, bis triphenylphosphoranylidene ammonium, triphenyl p,p,p-triphenylphosphine imidato-n phosphorus 1+ tetracarbonylcobaltate 1-, bis triphenylphosphine iminium chloride∼ppncl, ppn chloride, ppn cl, ksc491c1r, parent, triphenyl p,p,p-triphenylphosphine imidato phosphorus 1+ chloride
triphenyl-[(triphenyl-$l^{5}-phosphanylidene)amino]phosphanium;chloride

Specifications

21050-13-5
C36H30ClNP2
5 g
1,1,1-triphenyl-n-triphenylphosphoranylidene phosphoraniminium chloride, bis triphenylphosphoranylidene ammonium chloride, bis triphenylphosphoranylidene ammonium, triphenyl p,p,p-triphenylphosphine imidato-n phosphorus 1+ tetracarbonylcobaltate 1-, bis triphenylphosphine iminium chloride∼ppncl, ppn chloride, ppn cl, ksc491c1r, parent, triphenyl p,p,p-triphenylphosphine imidato phosphorus 1+ chloride
LVRCYPYRKNAAMX-UHFFFAOYSA-M
triphenyl-[(triphenyl-$l^{5}-phosphanylidene)amino]phosphanium;chloride
3036656
97%
(dry wt.), water <3%
274°C to 276°C
MFCD00151523
4097979
Moderately soluble in water.
[Cl-].C1=CC=C(C=C1)P(=[N+]=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
574.04
574.05
Bis(triphenylphosphoranylidene)ammonium chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 244-170-6

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only