missing translation for 'onlineSavingsMsg'
Learn More

Bis(trimethylsilyl)trifluoroacetamide, BSTFA, EMD-Millipore

$301.64 - $1225.61

Chemical Identifiers

CAS 25561-30-2
Molecular Formula C8H18F3NOSi2
Molecular Weight (g/mol) 257.403
InChI Key XCOBLONWWXQEBS-GHXNOFRVSA-N
Synonym 2,2,2-Trifluoro-N,O-bis(trimethylsilyl)acetamide
PubChem CID 9601896
IUPAC Name trimethylsilyl (1Z)-2,2,2-trifluoro-N-trimethylsilylethanimidate
SMILES C[Si](C)(C)N=C(C(F)(F)F)O[Si](C)(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
M1102550005
View Documents
Technidata
1102550005
5 mL Glass
Each for $301.64
Only null left
 
M1102550025
View Documents
Technidata
1102550025
25 mL Glass
Each for $1,225.61
Only null left
 
Description

Description

Specifications

Chemical Identifiers

25561-30-2
257.403
2,2,2-Trifluoro-N,O-bis(trimethylsilyl)acetamide
trimethylsilyl (1Z)-2,2,2-trifluoro-N-trimethylsilylethanimidate
C8H18F3NOSi2
XCOBLONWWXQEBS-GHXNOFRVSA-N
9601896
C[Si](C)(C)N=C(C(F)(F)F)O[Si](C)(C)C

Specifications

Glass
Density: 0.97g/cm3 (20°C)
2,2,2-Trifluoro-N,O-bis(trimethylsilyl)acetamide
C[Si](C)(C)N=C(C(F)(F)F)O[Si](C)(C)C
257.403
9601896
Bis(trimethylsilyl)trifluoroacetamide
25561-30-2
C8H18F3NOSi2
XCOBLONWWXQEBS-GHXNOFRVSA-N
trimethylsilyl (1Z)-2,2,2-trifluoro-N-trimethylsilylethanimidate
5 mL
257.4
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at 15° to 25°C

SDS
Documents

Documents