missing translation for 'onlineSavingsMsg'
Learn More

Bis(trimethylsilyl)telluride, 98%

Catalog No. AC291660050
Change view
Click to view available options
Quantity:
5 mL
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC291660050 5 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC291660050 Supplier Thermo Scientific Chemicals Supplier No. 291660050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4551-16-0 | C6H18Si2Te | 273.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4551-16-0
Molecular Formula C6H18Si2Te
Molecular Weight (g/mol) 273.97
MDL Number MFCD00143782
InChI Key VMDCDZDSJKQVBK-UHFFFAOYSA-N
Synonym bis-trimethylsilyl telluride, bis trimethylsilyl telluride, disilatellurane, hexamethyl, bis trimethylsilyl-telluride, trimethyl trimethylsilyltellanyl silane, et 2s,3s-2,3-epoxy-3-metpro, 1,1,1,3,3,3-hexamethyldisilatellurane #, disilatellurane,1,1,1,3,3,3-hexamethyl
PubChem CID 553023
IUPAC Name trimethyl(trimethylsilyltellanyl)silane
SMILES C[Si](C)(C)[Te][Si](C)(C)C

Specifications

Melting Point 13.5°C
Color Brown-Orange
Density 0.9700g/mL
Boiling Point 74°C (11.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula (Si(CH3)3)2Te
Quantity 5 mL
Specific Gravity 0.97
Solubility Information Solubility in water: .. Other solubilities: soluble in toluene,thf,hydrocarbons
Formula Weight 273.97
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Bis(trimethylsilyl)telluride, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement MOISTURE SENSITIVE

RUO – Research Use Only