missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(trimethylsilyl)sulfide, tech.
Description
Bis(trimethylsilyl)sulfide is used to convert oxides and chlorides into the corresponding sulfides. It has been used in the conversion of aldehydes and ketones to the corresponding thiones.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Colorless |
| Density | 0.851 g/mL |
| Boiling Point | 162°C to 166°C |
| Flash Point | 26°C (79°F) |
| Odor | Pungent |
| Packaging | Ampule |
| Refractive Index | 1.4598 |
| Linear Formula | [(CH3)3Si]2S |
| Quantity | 1 g |
| UN Number | UN1993 |
| Show More |
Safety and Handling
| Hazard Category | H226-H315-H319-H335 |
| Hazard Statement | GHS H Statement H226-H315-H319-H335 Flammable liquid and vapor. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c |
| DOTInformation | Hazard Class: 3; Packaging Group: III |
| EINECSNumber | 222-201-4 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only