Learn More
Bis(trimethylsilyl)sulfide, 95%
CAS: 3385-94-2 | C6H18SSi2 | 178.45 g/mol
$101.32 - $1126.37
Chemical Identifiers
| CAS | 3385-94-2 |
|---|---|
| Molecular Formula | C6H18SSi2 |
| Molecular Weight (g/mol) | 178.45 |
| MDL Number | MFCD00014851 |
| InChI Key | RLECCBFNWDXKPK-UHFFFAOYSA-N |
| Synonym | bis trimethylsilyl sulfide, hexamethyldisilthiane, hexamethyldisilathiane, disilathiane, hexamethyl, 1,1,1,3,3,3-hexamethyldisilathiane, disilthiane, hexamethyl, disilathiane, 1,1,1,3,3,3-hexamethyl, thiobis trimethylsilane, acmc-1ckj8 |
| PubChem CID | 76920 |
| IUPAC Name | trimethyl(trimethylsilylsulfanyl)silane |
| SMILES | C[Si](C)(C)S[Si](C)(C)C |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC367370010
|
Thermo Scientific Chemicals
367370010 |
1 g | Glass bottle |
Each for $101.32
|
|
||||
|
AC367370050
|
Thermo Scientific Chemicals
367370050 |
5 g | Glass bottle |
Each for $272.83
|
|
||||
|
AC367370250
|
Thermo Scientific Chemicals
367370250 |
25 g | Glass bottle |
Each for $1,126.37
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 3385-94-2 | |
| 178.45 | |
| RLECCBFNWDXKPK-UHFFFAOYSA-N | |
| 76920 | |
| C[Si](C)(C)S[Si](C)(C)C |
| C6H18SSi2 | |
| MFCD00014851 | |
| bis trimethylsilyl sulfide, hexamethyldisilthiane, hexamethyldisilathiane, disilathiane, hexamethyl, 1,1,1,3,3,3-hexamethyldisilathiane, disilthiane, hexamethyl, disilathiane, 1,1,1,3,3,3-hexamethyl, thiobis trimethylsilane, acmc-1ckj8 | |
| trimethyl(trimethylsilylsulfanyl)silane |
Specifications
| 3385-94-2 | |
| 0.9500g/mL | |
| 26°C | |
| 94% min. (GC) | |
| C6H18SSi2 | |
| S(Si(CH3)3)2 | |
| 1 g | |
| bis trimethylsilyl sulfide, hexamethyldisilthiane, hexamethyldisilathiane, disilathiane, hexamethyl, 1,1,1,3,3,3-hexamethyldisilathiane, disilthiane, hexamethyl, disilathiane, 1,1,1,3,3,3-hexamethyl, thiobis trimethylsilane, acmc-1ckj8 | |
| RLECCBFNWDXKPK-UHFFFAOYSA-N | |
| trimethyl(trimethylsilylsulfanyl)silane | |
| 76920 | |
| 95% | |
| Bis(trimethylsilyl)sulfide, 95% |
| Colorless to Yellow | |
| 163.0°C (170.0 mmHg) | |
| Authentic | |
| Glass bottle | |
| 1.4555 to 1.4585 | |
| MFCD00014851 | |
| 0.95 | |
| Solubility in water: .. Other solubilities: soluble in toluene,ethers,hydrocarbons | |
| C[Si](C)(C)S[Si](C)(C)C | |
| 178.45 | |
| 178.45 | |
| Liquid |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Flammable liquid and vapor.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 222-201-4
RUO – Research Use Only