missing translation for 'onlineSavingsMsg'
Learn More

Bis(trimethylsilyl)methylamine 95.0+%, TCI America™

Catalog No. B18261G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B18261G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B18261G Supplier TCI America Supplier No. B18261G
Only null left

Chemical Identifiers

CAS 134340-00-4
Molecular Formula C7H21NSi2
MDL Number MFCD00008262

Specifications

Color Yellow
Boiling Point 70°C
Quantity 1 g
UN Number 2920
Formula Weight 175.42
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Bis(trimethylsilyl)methylamine