CAS RN 134340-00-4
CAS RN 134340-00-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Bis(trimethylsilyl)methylamine 95.0+%, TCI America™
CAS: 134340-00-4 Molecular Formula: C7H21NSi2 MDL Number: MFCD00008262