missing translation for 'onlineSavingsMsg'
Learn More

Bis(trimethylsilyl)bromomethane 95.0+%, TCI America™

Catalog No. p-7119820
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B28741G 1 g
B28745G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B28741G Supplier TCI America Supplier No. B28741G
Only null left

Chemical Identifiers

CAS 29955-12-2
Molecular Formula C7H19BrSi2
Molecular Weight (g/mol) 239.30
MDL Number MFCD09038502
InChI Key MYXGYPFZVNSVCB-UHFFFAOYSA-N
Synonym Bis(trimethylsilyl)methyl Bromide
PubChem CID 11736575
IUPAC Name [bromo(trimethylsilyl)methyl]trimethylsilane
SMILES C[Si](C)(C)C(Br)[Si](C)(C)C

Specifications

Color Yellow
Quantity 1 g
UN Number 1760
Formula Weight 239.30
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Bis(trimethylsilyl)bromomethane
TSCA No
Recommended Storage Refrigerator