missing translation for 'onlineSavingsMsg'
Learn More

Bis(trimethylenedithio)tetrathiafulvalene 97.0+%, TCI America™

Catalog No. b1244100mg
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1244100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B1244100MG Supplier TCI America Supplier No. B1244100MG
Only null left

Chemical Identifiers

CAS 66946-49-4
Molecular Formula C12H12S8
Molecular Weight (g/mol) 412.71
MDL Number MFCD00060080
InChI Key LSZPCLOYNFOXST-UHFFFAOYSA-N
Synonym Bis(1,3-propylenedithio)tetrathiafulvalene, BPDT-TTF
PubChem CID 634371
IUPAC Name 2-{2H,5H,6H,7H-[1,3]dithiolo[4,5-b][1,4]dithiepin-2-ylidene}-2H,5H,6H,7H-[1,3]dithiolo[4,5-b][1,4]dithiepine
SMILES C1CSC2=C(SC(S2)=C2SC3=C(S2)SCCCS3)SC1

Specifications

Melting Point 250°C
Quantity 100 mg
Formula Weight 412.71
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Bis(trimethylenedithio)tetrathiafulvalene [Organic Electronic Material]