missing translation for 'onlineSavingsMsg'
Learn More

[Bis(trifluoroacetoxy)iodo]pentafluorobenzene 97.0+%, TCI America™

Catalog No. B16161G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B16161G 1 g
B16165G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B16161G Supplier TCI America Supplier No. B16161G

Chemical Identifiers

CAS 14353-88-9
Molecular Formula C10F11IO4
Molecular Weight (g/mol) 519.99
MDL Number MFCD00191612
InChI Key OQWAXRPJEPTTSZ-UHFFFAOYSA-N
Synonym Pentafluoro[bis(trifluoroacetoxy)iodo]benzene
PubChem CID 626316
IUPAC Name (2,3,4,5,6-pentafluorophenyl)[(2,2,2-trifluoroacetyl)oxy]-λ³-iodanyl 2,2,2-trifluoroacetate
SMILES FC1=C(F)C(F)=C([I](OC(=O)C(F)(F)F)OC(=O)C(F)(F)F)C(F)=C1F

Specifications

Melting Point 120°C
Color White
Quantity 1 g
UN Number 1759
Formula Weight 519.99
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material [Bis(trifluoroacetoxy)iodo]pentafluorobenzene