Learn More
Bis(tricyclohexylphosphine)palladium(0), 97% min
Air Sensitive | CAS: 33309-88-5 | C36H66P2Pd | 667.29 g/mol
Supplier: Thermo Scientific Chemicals 045792MD
Description
Bis(tricyclohexylphosphine)palladium(0) is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Bis(tricyclohexylphosphine)palladium(0) | |
| White | |
| MFCD01073796 | |
| Air and moisture sensitive | |
| Soluble in chloroform,toluene. Insoluble in alcohol and water. | |
| [Pd].C1CCC(CC1)P(C1CCCCC1)C1CCCCC1.C1CCC(CC1)P(C1CCCCC1)C1CCCCC1 | |
| 667.29 | |
| 667.29 |
| 33309-88-5 | |
| C36H66P2Pd | |
| 250 mg | |
| bis tricyclohexylphosphine palladium 0, bis tricyclohexylphosphine palladium o, bis tricyclohexylphosphine palladium, palladium, bis tricyclohexylphosphine, pd pcy3 2, palladium; tricyclohexylphosphane, palladium-tricyclohexylphosphine 1:2, bis tricyclohexylphosphine-palladium o | |
| JGBZTJWQMWZVNX-UHFFFAOYSA-N | |
| palladium;tricyclohexylphosphane | |
| 2734559 | |
| Crystalline |
Chemical Identifiers
| 33309-88-5 | |
| 667.29 | |
| JGBZTJWQMWZVNX-UHFFFAOYSA-N | |
| 2734559 | |
| [Pd].C1CCC(CC1)P(C1CCCCC1)C1CCCCC1.C1CCC(CC1)P(C1CCCCC1)C1CCCCC1 |
| C36H66P2Pd | |
| MFCD01073796 | |
| bis tricyclohexylphosphine palladium 0, bis tricyclohexylphosphine palladium o, bis tricyclohexylphosphine palladium, palladium, bis tricyclohexylphosphine, pd pcy3 2, palladium; tricyclohexylphosphane, palladium-tricyclohexylphosphine 1:2, bis tricyclohexylphosphine-palladium o | |
| palladium;tricyclohexylphosphane |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only