missing translation for 'onlineSavingsMsg'
Learn More

Bis(tricyclohexylphosphine)palladium(0), 97% min

Catalog No. p-4824626
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA45792MD 250 mg
AA4579203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA45792MD Supplier Thermo Scientific Chemicals Supplier No. 045792MD
Only null left

Air Sensitive | CAS: 33309-88-5 | C36H66P2Pd | 667.29 g/mol

Bis(tricyclohexylphosphine)palladium(0) is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 33309-88-5
Molecular Formula C36H66P2Pd
Molecular Weight (g/mol) 667.29
MDL Number MFCD01073796
InChI Key JGBZTJWQMWZVNX-UHFFFAOYSA-N
Synonym bis tricyclohexylphosphine palladium 0, bis tricyclohexylphosphine palladium o, bis tricyclohexylphosphine palladium, palladium, bis tricyclohexylphosphine, pd pcy3 2, palladium; tricyclohexylphosphane, palladium-tricyclohexylphosphine 1:2, bis tricyclohexylphosphine-palladium o
PubChem CID 2734559
IUPAC Name palladium;tricyclohexylphosphane
SMILES [Pd].C1CCC(CC1)P(C1CCCCC1)C1CCCCC1.C1CCC(CC1)P(C1CCCCC1)C1CCCCC1

Specifications

Color White
Quantity 250 mg
Sensitivity Air and moisture sensitive
Solubility Information Soluble in chloroform,toluene. Insoluble in alcohol and water.
Formula Weight 667.29
Physical Form Crystalline
Chemical Name or Material Bis(tricyclohexylphosphine)palladium(0)
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only